Calcium nitrate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
C4955-1KG C4955-500G
SIALC4955-1KGEA 336 EUR
SIALC4955-1KG SIALC4955-500G
Calcium nitrate tetrahydrate, Sigma-Aldrich®
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
Melting Pt: ∼42 °C
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III
Merck Index: 13,01687