Magnesium nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
203696-10G 203696-50G
SIAL203696-10GEA 68.4 EUR
SIAL203696-10G SIAL203696-50G
Magnesium nitrate hexahydrate, Sigma-Aldrich®
Magnesium nitrate hexahydrate
Formula: Mg(NO₃)₂·6H₂O
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 89 °C
Density: 1.63 g/cm³ (20 °C)
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00149779
CAS Number: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1,III
Merck Index: 13,05697