Strontium hydroxide octahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
463752-100G 463752-25G 463752-5G
SIAL463752-100GEA 804 EUR
SIAL463752-100G SIAL463752-25G SIAL463752-5G
Strontium hydroxide octahydrate, Sigma-Aldrich®
Strontium hydroxide octahydrate
Formula: H₂O₂Sr·8H₂O
MW: 265.76 g/mol
Melting Pt: 375 °C
Density: 1.9 g/cm³ (20 °C)
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,III