Magnesium sulphate monohydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
434183-1KG 434183-500G
SIAL434183-1KGEA 366 EUR
SIAL434183-1KG SIAL434183-500G
Magnesium sulphate monohydrate, Sigma-Aldrich®
Magnesium sulphate monohydrate
Formula: MgSO₄·1H₂O
Density: 2.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149787
CAS Number: 14168-73-1
EINECS: 231-298-2