Bole white (kaolin), Sigma-Aldrich®

Supplier: Merck

Synonyms: China Clay, Hydrated aluminum silicate, Clay

MQ200

18616-25KG 18616-5KG
SIAL18616-25KGEA 193 EUR
SIAL18616-25KG SIAL18616-5KG
Bole white (kaolin), Sigma-Aldrich®
Bole white (Kaolin)
Formula: Al₂SiO₇·2H₂O
MW: 258.16 g/mol
Melting Pt: 1760
Density: 0.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR