Halloysite nanoclay, Sigma-Aldrich®

Supplier: Merck

Synonyms: Kaolin clay

MQ100

685445-100G 685445-500G
SIAL685445-100GEA 86.6 EUR
SIAL685445-100G SIAL685445-500G
Halloysite nanoclay, Sigma-Aldrich®
Halloysite nanoclay
Formula: Al₂Si₂O₅(OH)₄·2H₂O
MW: 294.19 g/mol
Density: 0.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD11045913
CAS Number: 1332-58-7
EINECS: 310-194-1
REACH: exempt

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR