Tetraethylammonium chloride monohydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tetrylammonium chloride monohydrate

86605-250G 86605-50G
SIAL86605-250GEA 167 EUR
SIAL86605-250G SIAL86605-50G
Tetraethylammonium chloride monohydrate, Sigma-Aldrich®
Tetraethylammonium chloride monohydrate
Formula: C₈H₂₀ClN·1H₂O
MW: 183.72 g/mol
MDL Number: MFCD00149992
CAS Number: 68696-18-4
EINECS: 200-267-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR