Tetramethylammonium hydroxide pentahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
T7505-100G T7505-25G T7505-5G
SIALT7505-100GEA 0 EUR
SIALT7505-100G SIALT7505-25G SIALT7505-5G
Tetramethylammonium hydroxide pentahydrate, Sigma-Aldrich®
Tetramethylammonium hydroxide pentahydrate
Formula: C₄H₁₃NO·5H₂O
Melting Pt: 65…68 °C
Density: 1.829 g/cm³ (20 °C)
MDL Number: MFCD00149566
CAS Number: 10424-65-4
EINECS: 200-882-9
UN: 3423
ADR: 8,II