Ammonium molybdate(VI) ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate
43206.22 43206.14
43206.22EA 1220 EUR
43206.22 43206.14
Ammonium molybdate(VI) ≥99.99% (metals basis)
Ammonium molybdate(VI)
Purity: 99.99% CAS No.: 13106-76-8 Molecular Formula: H8MoN2O4 Formula Weight: 196.02 Physical Form: Powder/Lump Appearance: White Odour: Odourless MDL No.: MFCD00011596
Formula: H₈MoN₂O₄
MW: 196.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011596
CAS Number: 13106-76-8
EINECS: 236-031-3