Ammonium heptamolybdate tetrahydrate
Supplier: Roth Carl
Warning
Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate
3666.1 3666.2 3666.3
ROTH3666.1EA 108 EUR
ROTH3666.1 ROTH3666.2 ROTH3666.3
Ammonium heptamolybdate tetrahydrate
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536