Ammonium iron(II) sulphate hexahydrate
Supplier: Roth Carl
Warning
Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
X884.1 X884.2 X884.3
ROTHX884.1EA 23.5 EUR
ROTHX884.1 ROTHX884.2 ROTHX884.3
Ammonium iron(II) sulphate hexahydrate
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 100 °C
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8