Ammonium iron(II) sulphate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ferrous Ammonium sulphate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate
M-Clarity™ quality level = MQ200
203505-100G 203505-25G 203505-5G
SIAL203505-100GEA 483 EUR
SIAL203505-100G SIAL203505-25G SIAL203505-5G
Ammonium iron(II) sulphate hexahydrate, Sigma-Aldrich®
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 100 °C
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8