Cerium trichloride heptahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
228931-100G 228931-25G 228931-500G
SIAL228931-100GEA 113 EUR
SIAL228931-100G SIAL228931-25G SIAL228931-500G
Cerium trichloride heptahydrate, Sigma-Aldrich®
Cerium trichloride heptahydrate
Formula: CeCl₃·7H₂O
Melting Pt: 848 °C
Density: 3.97 g/cm³ (20 °C)
MDL Number: MFCD00149634
CAS Number: 18618-55-8
EINECS: 232-227-8
Merck Index: 12,02041