Erbium trichloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
289256-100G 289256-25G
SIAL289256-100GEA 363 EUR
SIAL289256-100G SIAL289256-25G
Erbium trichloride hexahydrate, Sigma-Aldrich®
Erbium trichloride hexahydrate
Formula: Cl₃Er·6H₂O
Melting Pt: 168 °C
MDL Number: MFCD00149693
CAS Number: 10025-75-9
EINECS: 233-385-0