Erbium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific
Warning
11304.14 11304.22
11304.14EA 123 EUR
11304.14 11304.22
Erbium trichloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium trichloride hydrate
Purity: 99.9% CAS No.: 19423-85-9 Molecular Formula: ErCl3.xH2O Formula Weight: 273.62(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic MDL No.: MFCD00010989
Formula: Cl₃Er·H₂O
MW: 291.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010989
CAS Number: 19423-85-9
EINECS: 233-385-0