Europium(III) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Europium(III) chloride hydrate, Europium trichloride hexahydrate
M-Clarity™ quality level = MQ200
203254-1G
203254-25G
203254-5G
SIAL203254-1GEA
40.7
EUR
SIAL203254-1G
SIAL203254-25G
SIAL203254-5G
Europium(III) chloride hexahydrate, Sigma-Aldrich®
Europium (III) chloride hexahydrate
Formula:
Cl₃Eu·6H₂O Melting Pt: 158 °C Density: 4.89 g/cm³ (20 °C) |
MDL Number:
MFCD00149701 CAS Number: 13759-92-7 EINECS: 233-040-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...