Erbium trinitrate pentahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Erbium(III) nitrate hydrate, Erbium (III) nitrate pentahydrate
M-Clarity™ quality level = MQ100
298166-100G 298166-25G
SIAL298166-100GEA 324 EUR
SIAL298166-100G SIAL298166-25G
Erbium trinitrate pentahydrate, Sigma-Aldrich®
Erbium trinitrate pentahydrate
Formula: ErN₃O₉·5H₂O
MW: 443.35 g/mol
MDL Number: MFCD00149691
CAS Number: 10031-51-3
EINECS: 233-436-7
UN: 1477
ADR: 5.1,III