Lanthanum(III) nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
331937-100G 331937-500G 331937-5G
SIAL331937-100GEA 205 EUR
SIAL331937-100G SIAL331937-500G SIAL331937-5G
Lanthanum(III) nitrate hexahydrate, Sigma-Aldrich®
Lanthanum (III) nitrate hexahydrate
Formula: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Boiling Pt: 126
Melting Pt: 65…67 °C
Density: 2.347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1 III
Merck Index: 14,05363