Neodymium trinitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
289175-100G 289175-25G
SIAL289175-100GEA 154 EUR
SIAL289175-100G SIAL289175-25G
Neodymium trinitrate hexahydrate, Sigma-Aldrich®
Neodymium trinitrate hexahydrate
Formula: N₃NdO₉·6H₂O
Melting Pt: ∼74 °C
Density: 2.265 g/cm³ (20 °C)
MDL Number: MFCD00149799
CAS Number: 16454-60-7
EINECS: 233-153-9
UN: 1477
ADR: 5.1,III