Samarium trinitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Samarium (III) nitrate hexahydrate, Samarium(III) nitrate hydrate
M-Clarity™ quality level = MQ100
298123-100G 298123-25G
SIAL298123-100GEA 374 EUR
SIAL298123-100G SIAL298123-25G
Samarium trinitrate hexahydrate, Sigma-Aldrich®
Samarium trinitrate hexahydrate
Formula: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Melting Pt: 78
Density: 2.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149857
CAS Number: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1 III