Terbium trinitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Terbium trinitrate 6-hydrate, Terbium(III) nitrate hexahydrate, Terbium(III) nitrate hexahydrate (1:3:6), Terbium (III) nitrate 6-hydrate, Terbium (III) nitrate hexahydrate
M-Clarity™ quality level = MQ100
217212-10G 217212-2G
SIAL217212-10GEA 535 EUR
SIAL217212-10G SIAL217212-2G
Terbium trinitrate hexahydrate, Sigma-Aldrich®
Terbium trinitrate hexahydrate
Formula: N₃O₉Tb·6H₂O
MW: 453.03
Melting Pt: 89.3
MDL Number: MFCD00149868
CAS Number: 13451-19-9
EINECS: 233-138-7