Copper(II) sulphate pentahydrate
Supplier: Hach
HACH1481866EA 34.5 EUR
HACH1481866
Copper(II) sulphate pentahydrate
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249,69 g/mol
Density: 2,284 g/cm³ (20 °C)
MDL Number: MFCD00149681
CAS Number: 7758-99-8
UN: 3077
ADR: 9,III
Merck Index: 13,02682