Copper(II) sulphate pentahydrate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL61245-500G-FEA 36.2 EUR
SIAL61245-500G-F SIAL61245-1KG-F SIAL61245-6X1KG-F
Copper(II) sulphate pentahydrate, Sigma-Aldrich®
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249,69 g/mol
Density: 2,284 g/cm³ (20 °C)
MDL Number: MFCD00149681
CAS Number: 7758-99-8
UN: 3077
ADR: 9,III
Merck Index: 13,02682

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR