Yttrium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Warning
11185.14 11185.22 11185.30
11185.14EA 67.7 EUR
11185.14 11185.22 11185.30
Yttrium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium (III) chloride hydrate
Purity: 99.99% CAS No.: 12741-05-8 Molecular Formula: Cl3Y.xH2O Formula Weight: 195.26(anhy) Physical Form: Crystalline Aggregates Density: 2.18 Appearance: White to pink Odour: Odourless Merck Reference: 14,10107 MDL No.: MFCD00149941
Formula: Cl₃Y·H₂O
MW: 213.28 g/mol
Density: 2.18
Storage Temperature: Ambient
MDL Number: MFCD00149941
CAS Number: 12741-05-8
EINECS: 233-801-0
Merck Index: 14,10107