Manganese(II) chloride tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate
M-Clarity™ quality level = MQ300
M8054-100G M8054-500G
SIALM8054-100GEA 104 EUR
SIALM8054-100G SIALM8054-500G
Manganese(II) chloride tetrahydrate, Sigma-Aldrich®
Manganese (II) chloride tetrahydrate
Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9 III
Merck Index: 13,05751