Manganese(II) sulphate monohydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIALM8179-500GEA 298 EUR
SIALM8179-500G SIALM8179-100G
Manganese(II) sulphate monohydrate, Sigma-Aldrich®
Manganese(II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169,02 g/mol
Melting Pt: 700 °C
Density: 2,95 g/cm³ (25 °C)
MDL Number: MFCD00149159
UN: 3077
ADR: 9,III
Merck Index: 13,05763