Nickel(II) bromide trihydrate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
72243-1KG
72243-250G
SIAL72243-1KGEA
632
EUR
SIAL72243-1KG
SIAL72243-250G
Nickel(II) bromide trihydrate, Sigma-Aldrich®
Nickel (II) bromide trihydrate
Formula:
Br₂Ni·3H₂O MW: 272.54 g/mol Melting Pt: 962…964 °C |
MDL Number:
MFCD00011141 CAS Number: 7789-49-3 EINECS: 236-665-0 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...