Iron(III) chloride hexahydrate
Supplier: Roth Carl
Danger
P742.2 P742.5
ROTHP742.2EA 133 EUR
ROTHP742.2 ROTHP742.5
Iron(III) chloride hexahydrate
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
Boiling Pt: 280…285 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM