Cobalt(II) nitrate hexahydrate 99%, pure
Supplier: Thermo Scientific
Danger
Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate
213091000 213095000 213090010 213090050
ACRO213091000EA 95.2 EUR
ACRO213091000 ACRO213095000 ACRO213090010 ACRO213090050
Cobalt(II) nitrate hexahydrate 99%, pure
Cobalt (II) nitrate hexahydrate
Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 55 °C
Density: 1,88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,II
Merck Index: 13,02469
Order Now
Specification Test Results
Appearance Pink to red to brown Adhering crystalline powder or crystals or chips
Titration Complexometric ≥98.5 %