Ruthenium(IV) oxide hydrate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
463760-1G 463760-5G
SIAL463760-1GEA 76.1 EUR
SIAL463760-1G SIAL463760-5G
Ruthenium(IV) oxide hydrate, Sigma-Aldrich®
Ruthenium (IV) oxide hydrate
Formula: O₂Ru·H₂O
MW: 151.08 g/mol
Density: 6.97 g/cm³ (20 °C)
MDL Number: MFCD00149846
CAS Number: 32740-79-7
EINECS: 234-840-6