Iron(II) sulphate heptahydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Iron vitriol, Ferrous sulphate heptahydrate
M-Clarity™ quality level = MQ200
F8263-1KG F8263-500G
SIALF8263-1KGEA 199 EUR
SIALF8263-1KG SIALF8263-500G
Iron(II) sulphate heptahydrate, Sigma-Aldrich®
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091