(R)-(-)-Linalool, Sigma-Aldrich®

Supplier: Merck

Synonyms: (-)-Linalool, (R)-3,7-Dimethylocta-1,6-dien-3-ol, (R)-(-)-3,7-Dimethyl-1,6-octadien-3-ol, L-Linalool

62139-100ML 62139-25ML
SIAL62139-100MLEA 152 EUR
SIAL62139-100ML SIAL62139-25ML
(R)-(-)-Linalool, Sigma-Aldrich®
(R)-(-)-Linalool
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00135469
CAS Number: 126-91-0
EINECS: 204-811-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR