N-Acetyl-L-cysteine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC
M-Clarity™ quality level = MQ200
A9165-25G
A9165-100G
A9165-5G
SIALA9165-25GEA
78.5
EUR
SIALA9165-25G
SIALA9165-100G
SIALA9165-5G
N-Acetyl-L-cysteine, Sigma-Aldrich®
N-Acetyl-L-cysteine
Formula:
C₅H₉NO₃S MW: 163.2 g/mol Boiling Pt: 371 Melting Pt: 106…108 °C Density: 1.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004880 CAS Number: 616-91-1 EINECS: 210-498-3 Merck Index: 13,00090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...