(+)-Dimethyl-L-tartrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Dimethyl (2R,3R)-2,3-dihydroxysuccinate, (R,R)-(+)-Tartaric acid dimethyl ester, (R,R)-(+)-Dimethyl tartrate, L(+)-Tartaric acid dimethyl ester, (+)-Dimethyl L-tartrate

M-Clarity™ quality level = MQ200

163457-100G 163457-25G
SIAL163457-100GEA 307 EUR
SIAL163457-100G SIAL163457-25G
(+)-Dimethyl-L-tartrate, Sigma-Aldrich®
(+)-Dimethyl L-tartrate
Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 58…61 °C
Density: 1.3403 g/cm³ (15 °C)
MDL Number: MFCD00064437
CAS Number: 608-68-4
EINECS: 210-166-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR