D-(-)-Arginine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: D-Arginine, (R)-2-Amino-5-guanidinopentanoic acid, H-D-Arg-OH, H-D-(-)-Arg-OH, (R)-(-)-Arginine, D-(-)-Arginine, D-2-Amino-5-guanidinovaleric acid
M-Clarity™ quality level = MQ200
A2646-1G A2646-250MG A2646-5G
SIALA2646-1GEA 103 EUR
SIALA2646-1G SIALA2646-250MG SIALA2646-5G
D-(-)-Arginine, Sigma-Aldrich®
D(-)-Arginine
Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00063116
CAS Number: 157-06-2
EINECS: 205-866-5