L(+)-Arginine Ph. Eur., USP, pure
Supplier: PanReac AppliChem
|
Warning
|
Synonyms:
H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
A1345.0500
A1345.1000
A1345.0500EA
122
EUR
A1345.0500
A1345.1000
L(+)-Arginine Ph. Eur., USP, pure
L(+)-Arginine
Formula:
C₆H₁₄N₄O₂ Boiling Pt: 368 °C (1013 hPa) Melting Pt: 238 °C Density: 1.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002635 CAS Number: 74-79-3 EINECS: 200-811-1 Merck Index: 13,00785 |