L(+)-Arginine ≥98%, Reagent Grade, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
M-Clarity™ quality level = MQ200
A5006-100G A5006-1KG A5006-500G
SIALA5006-100GEA 83.5 EUR
SIALA5006-100G SIALA5006-1KG SIALA5006-500G
L(+)-Arginine ≥98%, Reagent Grade, Sigma-Aldrich®
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785