L(+)-Arginine ≥99% FCC, FG, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
M-Clarity™ quality level = MQ300
W381918-10KG W381918-1KG W381918-5KG W381918-SAMPLE
SIALW381918-10KGEA 500 EUR
SIALW381918-10KG SIALW381918-1KG SIALW381918-5KG SIALW381918-SAMPLE
L(+)-Arginine ≥99% FCC, FG, Sigma-Aldrich®
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785