L-Arginine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride
MQ200
A5131-500G A5131-100G A5131-10G A5131-1KG A5131-25G A5131-25KG
SIALA5131-500GEA 285 EUR
SIALA5131-500G SIALA5131-100G SIALA5131-10G SIALA5131-1KG SIALA5131-25G SIALA5131-25KG
L-Arginine hydrochloride, Sigma-Aldrich®
L-Arginine hydrochloride
Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Melting Pt: 218
Density: 1.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785