L(+)-Cysteine, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

M-Clarity™ quality level = MQ200

C7352-10MG C7352-100G C7352-1KG C7352-25G
SIALC7352-10MGEA 32.6 EUR
SIALC7352-10MG SIALC7352-100G SIALC7352-1KG SIALC7352-25G
L(+)-Cysteine, Sigma-Aldrich®
L-Cysteine
Formula: HSCH₂CH(NH₂)CO₂H
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1.523 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
EINECS: 200-158-2
Merck Index: 13,02810

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR