L(+)-Lactic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid
M-Clarity™ quality level = MQ200
L1750-100G L1750-10G
SIALL1750-100GEA 700 EUR
SIALL1750-100G SIALL1750-10G
L(+)-Lactic acid, Sigma-Aldrich®
L(+)-Lactic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235
Melting Pt: ∼53 °C
Density: ∼1.385 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352