Sodium gluconate, Sigma-Aldrich®
Supplier: Merck
Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate
M-Clarity™ quality level = MQ200
S2054-100G S2054-1KG S2054-500G
SIALS2054-100GEA 40.7 EUR
SIALS2054-100G SIALS2054-1KG SIALS2054-500G
Sodium gluconate, Sigma-Aldrich®
Sodium gluconate
Formula: HOH₂C(CH(OH))₄COONa
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM