Calcium-D-gluconate monohydrate ≥98%, Sigma-Aldrich®
Supplier: Merck
Synonyms: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate
M-Clarity™ quality level = MQ200
G4625-1KG G4625-500G
SIALG4625-1KGEA 128 EUR
SIALG4625-1KG SIALG4625-500G
Calcium-D-gluconate monohydrate ≥98%, Sigma-Aldrich®
Calcium D-gluconate monohydrate
Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 178
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8