D-(-)-Glutamic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
M-Clarity™ quality level = MQ200
G1001-100G G1001-1G G1001-25G G1001-5G
SIALG1001-100GEA 1060 EUR
SIALG1001-100G SIALG1001-1G SIALG1001-25G SIALG1001-5G
D-(-)-Glutamic acid, Sigma-Aldrich®
D(-)-Glutamic acid
Formula: C₅H₉NO₄
MW: 147.13 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 213 °C
Density: 1.533 g/cm³ (20 °C)
MDL Number: MFCD00063112
CAS Number: 6893-26-1
EINECS: 230-000-8
Merck Index: 13,04482