L(+)-Aspartic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-(+)-Aminosuccinic acid (S)-Aminobutanedioic acid (S)-Aminosuccinic acid H-Asp-OH L-Asparaginic acid L(+)-Aspartic acid, (S)-(+)-Aspartic acid, L(+)-2-Amino-1,4-butanedioic acid, (S)-(+)-2-Amino-1,4-butanedioic acid, L-Aspartic acid, H-Asp-OH, L-α-Aminosuccinic acid, L-αlpha-Aminosuccinic acid, (S)-2-Aminosuccinic acid, Asp, D
M-Clarity™ quality level = MQ300
A7219-100G A7219-1KG
SIALA7219-100GEA 50.2 EUR
SIALA7219-100G SIALA7219-1KG
L(+)-Aspartic acid, Sigma-Aldrich®
L(+)-Aspartic acid
Formula: C₄H₇NO₄
MW: 133.1 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 251 °C
Density: 1.6613 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002616
CAS Number: 56-84-8
EINECS: 200-291-6
Merck Index: 13,00845