L(+)-Glutamic acid

Supplier: Molekula

MOLEM12307248EA 0 EUR
MOLEM12307248
L(+)-Glutamic acid
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
MW: 147,13 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1,538 g/cm³ (20 °C)
MDL Number: MFCD00002634
CAS Number: 56-86-0
https://si.vwr.com/stibo/web/std.lang.all/98/27/2299827.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR