L(+)-Glutamic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W328502-10KG-K W328502-1KG-K W328502-5KG-K
SIALW328502-10KG-KEA 510 EUR
SIALW328502-10KG-K SIALW328502-1KG-K SIALW328502-5KG-K
L(+)-Glutamic acid, Sigma-Aldrich®
L(+)-Glutamic acid
Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM