L(+)-Tartaric acid, BioXtra, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
M-Clarity™ quality level = MQ200
T1807-100G T1807-1KG T1807-500G
SIALT1807-100GEA 70.4 EUR
SIALT1807-100G SIALT1807-1KG SIALT1807-500G
L(+)-Tartaric acid, BioXtra, Sigma-Aldrich®
L(+)-Tartaric acid
Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399 °C (1013 hPa)
Melting Pt: 168…170 °C
Density: 1.759 g/cm³ (20 °C)
Flash Pt: 150 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM