D-(-)-Tartaric acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: unnatural tartaric acid, (2S,3S)-2,3-Dihydroxysuccinic acid, (S,S)-(-)-Tartaric acid, (S,S)-(-)-2,3-Dihydroxysuccinic acid
M-Clarity™ quality level = MQ200
95320-10G 95320-250G 95320-50G
SIAL95320-10GEA 40.4 EUR
SIAL95320-10G SIAL95320-250G SIAL95320-50G
D-(-)-Tartaric acid, Sigma-Aldrich®
D(-)-Tartaric acid
Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 166…169 °C
Density: 1.76 g/cm³ (20 °C)
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004238
CAS Number: 147-71-7
EINECS: 205-695-6
Merck Index: 12,09235