L(+)-Potassium sodium tartrate tetrahydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

M-Clarity™ quality level = MQ200

32312-1KG 32312-250G 32312-500G
SIAL32312-1KGEA 127 EUR
SIAL32312-1KG SIAL32312-250G SIAL32312-500G
L(+)-Potassium sodium tartrate tetrahydrate, Sigma-Aldrich®
L(+)-Potassium sodium tartrate tetrahydrate
Formula: KOOC(CHOH)₂COONa·4H₂O
Melting Pt: 70…80 °C
Density: ∼1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR